missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MURF1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518139
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine and canine based on sequence homology.
MURF1 Polyclonal antibody specifically detects MURF1 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
MURF1 | |
Polyclonal | |
Unconjugated | |
TRIM63 | |
E3 ubiquitin-protein ligase TRIM63; FLJ32380; IRF; iris ring finger protein; Murf; MURF 1; MuRF1; MuRF-1; MURF2; muscle ring finger protein 1; muscle specific ring finger protein 2; muscle-specific RING finger protein 1; OTTHUMP00000008701; RF1; ring finger protein 28; RING-type E3 ubiquitin transferase TRIM63; RNF 28; RNF28; SMRZ; striated muscle RING zinc finger protein; TRIM 63; Trim63; tripartite motif containing 63; tripartite motif containing 63, E3 ubiquitin protein ligase; tripartite motif protein 63; tripartite motif-containing 63; tripartite motif-containing protein 63 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
84676 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q969Q1 | |
TRIM63 | |
Synthetic peptide sequence (DYKSSLIQDGNPM) corresponding to the N-terminus amino acids of TRIM63 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |