Learn More
MSI1 Recombinant Polyclonal Antibody (13HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Marke: Invitrogen 710237
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with equine, mouse and rat based on sequence homology. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
MSI1 Recombinant Polyclonal antibody specifically detects MSI1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
MSI1 | |
Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
fb40b12; hypothetical protein LOC541389; m-Msi-1; ms1; msi1; msi1b; Msi1h; Musahi1; musashi homolog 1; Musashi homolog 1(Drosophila); musashi RNA binding protein 1; musashi RNA binding protein 1b; musashi RNA-binding protein 1; musashi1; musashi-1; RNA-binding protein Musashi homolog 1; RNA-binding protein Musashi homolog 1 L+35bp; RNA-binding protein Musashi homolog 1 Long; RNA-binding protein Musashi homolog 1b; wu:fb40b12; zgc:103751; zMsi1 | |
Recombinant protein corresponding to amino acids 280–362 of human Musashi-1 | |
100 μg | |
Primary | |
Human | |
Liquid |
Immunocytochemistry, Western Blot | |
13HCLC | |
Unconjugated | |
O43347 | |
Rabbit | |
Protein A | |
RUO | |
4440 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |