missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSI1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517464
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
MSI1 Polyclonal antibody specifically detects MSI1 in Human, Mouse, Rat, Zebrafish samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
MSI1 | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
O43347, Q61474, Q8K3P4 | |
MSI1 | |
Synthetic peptide corresponding to residues of human Musashi | |
100 μL | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Antibody | |
IgG |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
MSI1 | |
fb40b12; hypothetical protein LOC541389; m-Msi-1; ms1; msi1; msi1b; Msi1h; Musahi1; musashi homolog 1; Musashi homolog 1(Drosophila); musashi RNA binding protein 1; musashi RNA binding protein 1b; musashi RNA-binding protein 1; musashi1; musashi-1; RNA-binding protein Musashi homolog 1; RNA-binding protein Musashi homolog 1 L+35bp; RNA-binding protein Musashi homolog 1 Long; RNA-binding protein Musashi homolog 1b; wu:fb40b12; zgc:103751; zMsi1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17690, 259272, 4440, 541389 | |
-20°C | |
Liquid |