Learn More
MBNL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529825
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, HeLa, Jurkat. Predicted reactivity: Mouse (98%), Rat (99%), Chicken (97%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MBNL1 Polyclonal antibody specifically detects MBNL1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
MBNL1 | |
Polyclonal | |
Unconjugated | |
MBNL1 | |
DKFZp686P06174; EXP; EXP35; EXP40; EXP42; KIAA0428; MBNL; MBNL1; mKIAA0428; muscleblind like splicing regulator 1; muscleblind-like 1; muscleblind-like 1 (Drosophila); muscleblind-like protein 1; muscleblind-like splicing regulator 1; triplet-expansion RNA-binding protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4154 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.45 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NR56 | |
MBNL1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 259 of Human Muscleblind-like 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |