missing translation for 'onlineSavingsMsg'
Learn More

MBNL1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA519739

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11347683

  • 469.00 € / 100 Mikrogramm
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Heat mediated antigen retrieval recommended prior to tissue staining. This antibody is predicted to react with mouse and rat based on sequence homology.

MBNL1 Polyclonal antibody specifically detects MBNL1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

MBNL1
Polyclonal
Unconjugated
MBNL1
DKFZp686P06174; EXP; EXP35; EXP40; EXP42; KIAA0428; MBNL; MBNL1; mKIAA0428; muscleblind like splicing regulator 1; muscleblind-like 1; muscleblind-like 1 (Drosophila); muscleblind-like protein 1; muscleblind-like splicing regulator 1; triplet-expansion RNA-binding protein
Rabbit
Antigen affinity chromatography
RUO
4154
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.02% sodium azide; pH 7.4
Q9NR56
MBNL1
Synthetic peptide conjugated to KLH derived from within residues 250 - 350 of Human Muscleblind-like 1.
100 μg
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote