Learn More
MUSK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527728
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1. Predicted reactivity: Mouse (97%), Rat (98%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MUSK Polyclonal antibody specifically detects MUSK in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MUSK | |
Polyclonal | |
Unconjugated | |
MUSK | |
CMS9; FADS; Mdk4; MGC126323; MGC126324; Mlk; muscle associated receptor tyrosine kinase; muscle localized kinase 2; muscle specific kinase (neural fold/somite kinase 1); muscle, skeletal receptor tyrosine protein kinase; Muscle, skeletal receptor tyrosine-protein kinase; muscle, skeletal, receptor tyrosine kinase; muscle-specific kinase receptor; muscle-specific protein kinase secretory isoform; muscle-specific tyrosine kinase receptor MuSK; muscle-specific tyrosine protein kinase receptor; Muscle-specific tyrosine-protein kinase receptor; MuSK; Nsk1; Nsk2; Nsk3; RP23-170A3.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4593 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O15146 | |
MUSK | |
Recombinant fragment corresponding to a region within amino acids 1 and 174 of Human MUSK | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |