missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUTYH Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527855
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, NIH-3T3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MUTYH Polyclonal antibody specifically detects MUTYH in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
MUTYH | |
Polyclonal | |
Unconjugated | |
MUTYH | |
5730495A01Rik; A/G-specific adenine DNA glycosylase; adenine DNA glycosylase; adenine-DNA glycosylase; hMYH; MGC4416; mMYH; mutY DNA glycosylase; mutY homolog; mutY homolog (E. coli); mutY homolog alpha; Mutyh; Mutyha; Mutyhalpha; Mutyhb; Mutyhbeta; Mutyhc; mutY-like protein; Myh; OTTHUMP00000009098; OTTHUMP00000009102; rMYH; RP4-534D1.2; the full-length form type A, 5' region | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4595, 70603 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.16 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q99P21, Q9UIF7 | |
MUTYH | |
Recombinant fragment corresponding to a region within amino acids 257 and 546 of Human MUTYH | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |