Learn More
Netrin-4 Rat anti-Human, Mouse, Clone: 299612, R&D Systems™
Rat Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB1132
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418385
  • 278.53 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 25-04-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Netrin-4 Monoclonal antibody MAB1132 Specifically detects Netrin-4 in Human, Mouse samples. It is validated for Western Blotting
Spezifikation
Spezifikation
Netrin-4
299612
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
NTN4
Mouse myeloma cell line NS0-derived recombinant mouse Netrin-4 Gly19-Val628 Accession # Q9JI33
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
59277
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9JI33
beta-netrin, hepar-derived netrin-like protein, netrin 4, Netrin4, NTN4, PRO3091
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human and mouse Netrin-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) Netrin-1, rmNetrin-G1a, rmNetrin-G2a, recombinant human Netrin-4, recombinant chicken (rch) Netrin-1, or rchNetrin-2 is observed.
Human, Mouse