missing translation for 'onlineSavingsMsg'
Learn More

Nope Rat anti-Mouse, Clone: 261123, R&D Systems™

Rat Monoclonal Antibody

Marke:  RD Systems (Bio-Techne) MAB1394

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13418605

  • 278.53 € / 100 Mikrogramm


Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Nope Monoclonal antibody MAB1394 Specifically detects Nope in Mouse samples. It is validated for Western Blotting
Spezifikation

Spezifikation

Nope
261123
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IGDCC4
Mouse myeloma cell line NS0-derived recombinant mouse Nope, Gly22-His953, Accession # Q9EQS9
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
57722
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9EQS9
DDM36, hDDM36, IGDCC4, immunoglobulin superfamily DCC subclass member 4, immunoglobulin superfamily, DCC subclass, member 4, KIAA1628, likely ortholog of mouse neighbor of Punc E11, LOC57722, Neighbor of punc e11, NOPEFLJ42051, Protein DDM36
Rat
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects mouse Nope in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse (rm) DCC, rmNeogenin, and rmPunc is observed.
Mouse
Videos

For Research Use Only