Learn More
Notch-2 Mouse anti-Rat, Clone: 199509, R&D Systems™
Mouse Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB1190
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418435
  • 377.53 € / 500 Mikrogramm
Voraussichtlicher Liefertermin 25-02-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Notch-2 Monoclonal antibody MAB1190 Specifically detects Notch-2 in Rat samples. It is validated for Western Blotting
Spezifikation
Spezifikation
Notch-2
199509
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
NOTCH2
S. frugiperda insect ovarian cell line Sf 21-derived recombinant rat Notch-2, Leu26-Glu492, Accession # Q9QW30
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4853
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9QW30
AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2, Notch2
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects rat Notch-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant rat Notch-1 or recombinant mouse Notch-3 is observed.
Rat