Learn More
O-linked N-acetylglucosamine (O-GlcNAc) Monoclonal Antibody (18B10.C7), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA1038
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
MA1-038 has been successfully used in Western blot and immunoprecipitation applications with human samples. In Western blot, MA1-038 detects proteins with O-GlcNAc post-translational modification, and therefore will detect multiple specific bands.
O-linked N-acetylglucosamine (O-GlcNAc) Monoclonal antibody specifically detects O-linked N-acetylglucosamine (O-GlcNAc) in Human samples. It is validated for Immunoprecipitation, Western Blot.Spezifikation
O-linked N-acetylglucosamine (O-GlcNAc) | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
O15294 | |
OGT | |
SANM-conjugated linear peptide corresponding to human O-GlcNAc. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunoprecipitation, Western Blot | |
18B10.C7 | |
Unconjugated | |
OGT | |
GlcNAc; HINCUT-1; HRNT1; O-GLCNAC; O-GlcNAc transferase p110 subunit; O-GlcNAc transferase subunit p110; OGT; O-linked N-acetylglucosamine (GlcNAc) transferase; O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase); O-linked N-acetylglucosamine transferase 110 kDa subunit; UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase; UDP-N-acetylglucosamine--peptide N-acetylglucosaminyltransferase 110 kDa subunit; uridinediphospho-N-acetylglucosamine:polypeptide beta-N-acetylglucosaminyl transferase | |
Mouse | |
Protein A | |
RUO | |
8473 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |