missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OAT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA535370
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
OAT Polyclonal antibody specifically detects OAT in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
OAT | |
Polyclonal | |
Unconjugated | |
OAT | |
AI194874; GACR; gyrate atrophy; HOGA; OAT; OATASE; OKT; ornithine aminotransferase; ornithine aminotransferase (gyrate atrophy); Ornithine aminotransferase, hepatic form; Ornithine aminotransferase, mitochondrial; Ornithine aminotransferase, renal form; Ornithine delta-aminotransferase; ornithine-delta-aminotransferase; ornithine--oxo-acid aminotransferase; ornithine-oxo-acid aminotransferase; rOAT; testicular tissue protein Li 130 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
18242, 4942, 64313 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P04181, P04182, P29758 | |
OAT | |
KLH conjugated synthetic peptide between 27-55 amino acids from the N-terminal region of human OAT | |
400 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |