missing translation for 'onlineSavingsMsg'
Learn More

OATP2 Monoclonal Antibody (ESL), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA3934

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11585812

  • 483.00 € / 100 Mikroliter
Verfügbar ab: 15-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

MA3-934 detects OATP from human tissue and cells. This antibody does not detect dog, mouse or rat protein. MA3-934 shows no cross-reactivity to other OATP isoforms. MA3-934 has been successfully used in immunofluorescence, western blotting, and immunoprecipitation procedures. Immunofluorescence staining of OATP2 in human liver samples results in a pattern consistent with plasma membrane staining. The MA3-934 immunogen is a synthetic peptide corresponding to residues E(671) S L N K N K H F V P S A G A D S E T H C(691) of human OATP2.

OATP2 Monoclonal antibody specifically detects OATP2 in Human samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Spezifikation

Spezifikation

OATP2
Monoclonal
Conc. Not Determined
tissue culture supernatant with 0.09% sodium azide
Q9Y6L6
SLCO1B1
Synthetic peptide corresponding to residues E(671) S L N K N K H F V P S A G A D S E T H C(691) of human OATP2.
RUO
10599
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunocytochemistry, Immunoprecipitation, Western Blot
ESL
Unconjugated
SLCO1B1
HBLRR; liver-specific organic anion transporter 1; LST1; LST-1; OATP1B1; OATP2; OATP-2; OATPC; OATP-C; SLC01B1; SLC21A6; SLCO1B1; Sodium-independent organic anion-transporting polypeptide 2; solute carrier family 21 (organic anion transporter), member 6; Solute carrier family 21 member 6; Solute carrier organic anion transporter family member 1B1; solute carrier organic anion transporter family, member 1B1
Mouse
100 μL
Primary
Human
Antibody
IgM
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote