Learn More
OATP2/OATP8 Monoclonal Antibody (MDQ), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA3935
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA3-935 detects OATP8 from human tissue and cells. This antibody does not detect dog, mouse or rat protein. MA3-935 also detects the OATP2 isoform. MA3-935 has been successfully used in immunohistochemistry, immunofluorescence and immunoprecipitation procedures. Immunofluorescence staining of OATP8 in human liver samples results in a pattern consistent with plasma membrane staining. The MA3-935 immunogen is a synthetic peptide corresponding ot residues M(1) D Q N Q H L N K T A E A Q P S E N K K T R Y(23) C of human OATP2.
OATP2/OATP8 Monoclonal antibody specifically detects OATP2/OATP8 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Spezifikation
OATP2/OATP8 | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.09% sodium azide | |
Q9NPD5, Q9Y6L6 | |
SLCO1B1, SLCO1B3 | |
Synthetic peptide corresponding to residues M(1) D Q N Q H L N K T A E A Q P S E N K K T R Y(23) C of human OATP2. | |
RUO | |
10599, 28234 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation | |
MDQ | |
Unconjugated | |
SLCO1B1 | |
HBLRR; liver-specific organic anion transporter 1; Liver-specific organic anion transporter 2; liver-specific organic anion transporter 3TM13; LST1; LST-1; LST2; LST-2; LST3; LST-3TM13; OATP1B1; OATP1B3; OATP2; OATP-2; OATP8; OATP-8; OATPC; OATP-C; Organic anion transporter 8; organic anion transporter LST-3c; organic anion-transporting polypeptide 8; SLC21A6; SLC21A8; SLCO1B1; SLCO1B3; Sodium-independent organic anion-transporting polypeptide 2; solute carrier family 21 (organic anion transporter), member 6; solute carrier family 21 (organic anion transporter), member 8; Solute carrier family 21 member 6; Solute carrier family 21 member 8; Solute carrier organic anion transporter family member 1B1; solute carrier organic anion transporter family member 1B3; solute carrier organic anion transporter family, member 1B1; solute carrier organic anion transporter family, member 1B3 | |
Mouse | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |