Learn More
OPCML Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA547392
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) KILON and rhNeurotrimin is observed and less than 1% cross-reactivity with rhALCAM, rhBCAM, rhEpCAM and rhNCAM is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
OPCML Polyclonal antibody specifically detects OPCML in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot.Spezifikation
OPCML | |
Polyclonal | |
Unconjugated | |
OPCML | |
2900075O15Rik; 3732419F12; AI844366; B930023M13Rik; C230027C17; Gm181; IgLON family member 1; IGLON1; OBCAM; OPCM; OPCML; opioid binding cell adhesion molecule; opioid binding protein/cell adhesion molecule like; opioid binding protein/cell adhesion molecule-like; opioid binding protein/cell adhesion molecule-like preprotein; opioid-binding cell adhesion molecule; opioid-binding protein/cell adhesion molecule; opioid-binding protein/cell adhesion molecule-like | |
Goat | |
Antigen affinity chromatography | |
RUO | |
4978 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q14982 | |
OPCML | |
Mouse myeloma cell line NS0-derived recombinant human OBCAM/OPCML Gly28-Asn322 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |