missing translation for 'onlineSavingsMsg'
Learn More

Occludin Recombinant Rabbit Monoclonal Antibody (6H10L9), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Marke:  Invitrogen 701161

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15367064

  • 456.00 € / 100 Mikrogramm
Verfügbar ab: 30-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.


Beschreibung

Beschreibung

This antibody is predicted to react with equine, mouse, rabbit and rat based on sequence homology. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Occludin Recombinant Monoclonal antibody specifically detects Occludin in Human samples. It is validated for Immunocytochemistry, Western Blot.
Spezifikation

Spezifikation

Occludin
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide
AI503564; BLCPMG; Occludin; occludin 1B; Ocl; Ocln; phosphatase 1, regulatory subunit 115; PPP1R115; tight junction protein occludin; tight junction structural protein
Peptide corresponding to amino acids 387–403 of human occludin
100 μg
Primary
Human
Liquid
Immunocytochemistry, Western Blot
6H10L9
Unconjugated
Q16625
Rabbit
Protein A
RUO
100506658
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote