missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Occludin Monoclonal Antibody (OC-3F10), HRP, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen 331520
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
Occludin Monoclonal antibody specifically detects Occludin in Canine, Human, Mouse, Rat samples. It is validated for ELISA, Western Blot.Spezifikation
Occludin | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol, 4mg/mL BSA and 0.19% Kathon™ CG; pH 7.4 | |
Q16625, Q28269, Q61146, Q6P6T5 | |
OCLN | |
GST fusion protein consisting of the C-terminal region (∽150aa) of human occludin. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
OC-3F10 | |
HRP | |
OCLN | |
AI503564; BLCPMG; Occludin; occludin 1B; Ocl; Ocln; phosphatase 1, regulatory subunit 115; PPP1R115; tight junction protein occludin; tight junction structural protein | |
Mouse | |
Protein A | |
RUO | |
100506658, 18260, 403844, 83497 | |
4° C | |
Liquid |