Learn More
Occludin Recombinant Polyclonal Antibody (6HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Marke: Invitrogen 710192
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
Occludin Recombinant Polyclonal antibody specifically detects Occludin in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Occludin | |
Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
AI503564; BLCPMG; Occludin; occludin 1B; Ocl; Ocln; phosphatase 1, regulatory subunit 115; PPP1R115; tight junction protein occludin; tight junction structural protein | |
Peptide corresponding to amino acids 387-403 of human occuludin | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
6HCLC | |
Unconjugated | |
Q16625 | |
Rabbit | |
Protein A | |
RUO | |
100506658 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |