missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Occludin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530230
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Occludin Polyclonal antibody specifically detects Occludin in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Occludin | |
Polyclonal | |
Unconjugated | |
OCLN | |
AI503564; BLCPMG; Occludin; occludin 1B; Ocl; Ocln; phosphatase 1, regulatory subunit 115; PPP1R115; tight junction protein occludin; tight junction structural protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100506658, 83497 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.98 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q16625, Q6P6T5 | |
OCLN | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 522 of Human Occludin | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |