Learn More
OCT1 (POU2F1) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528209
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (89%), Rat (94%), Xenopus laevis (81%), Pig (97%), Chicken (94%), Chimpanzee (99%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
OCT1 (POU2F1) Polyclonal antibody specifically detects OCT1 (POU2F1) in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
OCT1 (POU2F1) | |
Polyclonal | |
Unconjugated | |
POU2F1 | |
2810482H01Rik; NF-A1; OCT1; oct-1; oct-1B; octamer binding protein 1; octamer-binding protein 1; Octamer-binding transcription factor 1; Octamer-binding transcription factor-1; Otf1; OTF-1; POU class 2 homeobox 1; POU domain, class 2, transcription factor 1; Pou2f1; ubiquitous transcription factor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
171068, 5451 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.49 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P14859, P31503 | |
POU2F1 | |
Recombinant protein fragment corresponding to a region within amino acids 442 and 755 of Human OCT1 | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |