missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT6 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA537883
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
This antibody is predicted to react with mouse, rat, dog and cow based on sequence homology.
OCT6 Polyclonal antibody specifically detects OCT6 in Human samples. It is validated for Western Blot.Spezifikation
OCT6 | |
Polyclonal | |
Unconjugated | |
POU3F1 | |
6-Oct; LOW QUALITY PROTEIN: POU domain, class 3, transcription factor 1; Oct6; Oct-6; octamer binding factor oct6; octamer-binding protein 6; Octamer-binding transcription factor 6; octoamer transcription factor-6; octomer-6; Otf6; OTF-6; POU class 3 homeobox 1; POU domain transcription factor SCIP; POU domain, class 3, transcription factor 1; POU3F1; Scip; suppressed cAMP-inducible POU; Test1; Testes-1; Tst1; Tst-1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
5453 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q03052 | |
POU3F1 | |
Peptide with sequence C-HHALHEDGHEAQLE, from the internal region of POU3F1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |