missing translation for 'onlineSavingsMsg'
Learn More

OMP Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Marke:  Invitrogen OSG00001W

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11520483

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

The peptide is homologous with the corresponding sequence derived from OMP protein in mouse. Reconstitute in 100 μL of sterile water.

Olfactory marker protein (OMP) is an abundant, 19-kDa, cytosolic protein that is almost exclusively expressed in mature, functioning, olfactory neurons but not in the neural precursor basal cells. The OMP gene structure and protein sequence are highly conserved between mouse, rat and human. Its tissue-specific expression in the receptor cells together with the results of the mouse knockout studies show that OMP represents a novel modulatory component of the odor detection/signal transduction cascade. May act as a modulator of the olfactory signal-transduction cascade. Interacts with BEX1 and BEX2. Uniquely associated with mature olfactory receptor neurons. Belongs to the olfactory marker protein family.
Spezifikation

Spezifikation

OMP
Polyclonal
Omp
Olfactory neuronal-specific protein
Goat
100 μL
Primary
Mouse, Rat
Antibody
IgG
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Q64288, P08523
OMP
Synthetic peptide from rat Olfactory Marker Protein.
RUO
18378, 24612
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote