Learn More
OMP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke:  Invitrogen OSR00001W
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 11594883
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Beschreibung
Beschreibung
In IHC applications, Zamboni's or 4% PFA fixatives are recommended. Reconstitute in 100 μL of sterile water.
Olfactory marker protein (OMP) is an abundant, 19-kDa, cytosolic protein that is almost exclusively expressed in mature, functioning, olfactory neurons but not in the neural precursor basal cells. The OMP gene structure and protein sequence are highly conserved between mouse, rat and human. Its tissue-specific expression in the receptor cells together with the results of the mouse knockout studies show that OMP represents a novel modulatory component of the odor detection/signal transduction cascade. May act as a modulator of the olfactory signal-transduction cascade. Interacts with BEX1 and BEX2. Uniquely associated with mature olfactory receptor neurons. Belongs to the olfactory marker protein family.
Spezifikation
Spezifikation
OMP
Polyclonal
Omp
Olfactory neuronal-specific protein
Rabbit
100 μL
Primary
Mouse, Rat
Antibody
IgG
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Q64288, P08523
OMP
A synthetic peptide from rat Olfactory Marker Protein
RUO
18378, 24612
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized