missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OMP Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen OSS00003W
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
In IHC applications, Zamboni's or 4% PFA fixatives are recommended. Reconstitute in 100 uL of sterile water.
Olfactory marker protein (OMP) is an abundant, 19-kDa, cytosolic protein that is almost exclusively expressed in mature, functioning, olfactory neurons but not in the neural precursor basal cells. The OMP gene structure and protein sequence are highly conserved between mouse, rat and human. Its tissue-specific expression in the receptor cells together with the results of the mouse knockout studies show that OMP represents a novel modulatory component of the odor detection/signal transduction cascade. May act as a modulator of the olfactory signal-transduction cascade. Interacts with BEX1 and BEX2. Uniquely associated with mature olfactory receptor neurons. Belongs to the olfactory marker protein family.Spezifikation
OMP | |
Polyclonal | |
OMP | |
Olfactory neuronal-specific protein | |
Sheep | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
P47874 | |
OMP | |
Synthetic peptide from human Olfactory Marker Protein. | |
RUO | |
4975 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |