missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olfr2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSR00013W
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material. OSR00013W can be used with blocking peptide GSM00005P.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.Spezifikation
Olfr2 | |
Polyclonal | |
Olfr2 | |
OLFR2 | |
Synthetic peptide from mouse olfactory receptor family 103, subfamily 15. | |
RUO | |
18317 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Olfactory receptor Olfr2, Olfactory receptor MOR103-15, I7 olfactory receptor, Olfactory receptor I7. ORL431, ORL535, Olfr41. GENE NAME: Olfr2 | |
Rabbit | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |