missing translation for 'onlineSavingsMsg'
Learn More

Olfr544 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen OSR00016W

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11514903

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material. OSR00016W can be used with blocking peptide GSM00020P.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Spezifikation

Spezifikation

Olfr544
Polyclonal
Olfr544
OLFR544
A synthetic peptidefrom mouse olfactory receptor family 42, subfamily 3 (MOR42-3) conjugated to an immunogenic carrier protein was used as the antigen
RUO
257926
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Immunohistochemistry, Western Blot
Unconjugated
Odorant receptor S6, Olfactory receptor 544, Olfactory receptor Olfr544, ORL471, S79, ORL463. GENE NAME: Olfr544.
Rabbit
100 μL
Primary
Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote