missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olfr56 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSR00014W
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Glycerol (1:1) may be added for additional stability. Reconstitute in 100 uL of sterile water. OSR00014W can be used with blocking peptide GSM00014P.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals.Spezifikation
Olfr56 | |
Polyclonal | |
Olfr56 | |
Olfactory receptor Olfr56, ORL436, G7. GENE NAME: Olfr56 | |
Rabbit | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q8VGD6 | |
OLFR56 | |
Synthetic peptide from mouse olfactory receptor family 276, subfamily 1. | |
RUO | |
18356 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |