missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olfr56 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSR00031G
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 500 μL of distilled water. Centrifuge to remove any insoluble material.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.Spezifikation
Olfr56 | |
Polyclonal | |
Olfr56 | |
Olfactory receptor Olfr56, ORL436, G7. GENE NAME: Olfr56 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
18356 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q8VGD6 | |
OLFR56 | |
Synthetic peptide from mouse olfactory receptor family 276, subfamily 1. | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |