missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olfr571 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSR00194W
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome.Spezifikation
Olfr571 | |
Polyclonal | |
Olfr571 | |
OLFR571 | |
Synthetic peptide from mouse olfactory receptor family 21, subfamily 1. | |
RUO | |
259089 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Olfactory receptor MOR21-1, olfactory receptor 571, Olfr571 | |
Rabbit | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |