missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olfr642 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSR00027G
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 500 μL of distilled water. Centrifuge to remove any insoluble material.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.Spezifikation
Olfr642 | |
Polyclonal | |
Olfr642 | |
OLFR642 | |
Synthetic peptide from mouse olfactory receptor family 13, subfamily 6, 2 and 5. | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Olfactory receptor S85, Olfactory receptor 642, Olfactory receptor Olfr642, Olfactory receptor MOR13-6, S85, ORL473, Olfr642, MOR13-2, Olfr, MOR13-6, MOR13-2, MOR13-5 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
258326 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |