missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR11H4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSR00008W
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material.
Putative odorant receptor. Multi-pass membrane protein. Belongs to the G-protein coupled receptor 1 family.Spezifikation
OR11H4 | |
Polyclonal | |
OR11H4 | |
Olfactory receptor OR14-36. GENE NAME: OR11H4 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
Q8NGC9 | |
OR11H4 | |
A synthetic peptide from c-terminal region of human olfactory receptor | |
RUO | |
390442 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |