missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OLIG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA521613
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Molt-4. Predicted reactivity: Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
OLIG1 Polyclonal antibody specifically detects OLIG1 in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.Spezifikation
OLIG1 | |
Polyclonal | |
Unconjugated | |
OLIG1 | |
AI836478; AW494459; basic domain, helix-loop-helix protein, class B, 6; BHLHB6; bHLHb7; bHLHe20; bHLHe21; class B basic helix-loop-helix protein 6; Class E basic helix-loop-helix protein 21; LOW QUALITY PROTEIN: oligodendrocyte transcription factor 1; Olg1; Olg-1; Olg-1 bHLH protein; OLIG1; olig2; olig3; Oligo1; Oligo2 antibody; oligodendrocyte lineage transcription factor 1; oligodendrocyte transcription factor 1; Oligodendrocyte-specific bHLH transcription factor 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116448, 60394 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
1 mg/mL | |
PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8TAK6, Q9WUQ3 | |
OLIG1 | |
Synthetic peptide corresponding to a region within amino acids 193 and 271 of OLIG1 (Uniprot ID#Q8TAK6) | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |