missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPA1 Monoclonal Antibody (1E8-1D9), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516149
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
In Western blot, multiple protein isoforms can be seen at approximately 90, 80 and 65 kDa.
OPA1 Monoclonal antibody specifically detects OPA1 in Bovine, Hamster, Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
OPA1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O60313, P58281, Q2TA68 | |
OPA1 | |
Human OPA1. | |
100 μL | |
Primary | |
Bovine, Hamster, Human, Mouse, Rat | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1E8-1D9 | |
Unconjugated | |
OPA1 | |
1200011N24Rik; AI225888; AI847218; BERHS; Dynamin-like 120 kDa protein, form S1; Dynamin-like 120 kDa protein, mitochondrial; dynamin-like guanosine triphosphatase; EMBL:AAI11072.1}; hypothetical protein; KIAA0567; Large GTP-binding protein; largeG; lilr3; MGM1; mitochondrial dynamin-like GTPase; mitochondrial OPA1; mKIAA0567; MTDPS14; NPG; NTG; OPA1; opa1 {ECO:0000312; OPA1 mitochondrial dynamin like GTPase; OPA1, mitochondrial dynamin like GTPase; optic atrophy 1; optic atrophy 1 (autosomal dominant); optic atrophy 1 homolog; optic atrophy 1-like protein; optic atrophy protein 1; Optic atrophy protein 1 homolog; RCJMB04_1m16; RN protein | |
Mouse | |
Protein G | |
RUO | |
100751873, 171116, 4976, 524142, 74143 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |