missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPRD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533468
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Dog, Elephant, Panda, Horse, Rabbit (100%).
OPRD1 Polyclonal antibody specifically detects OPRD1 in Canine, Human, Non-human Primate, Rabbit samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
OPRD1 | |
Polyclonal | |
Unconjugated | |
OPRD1 | |
delta 1 opioid receptor; delta opioid receptor 1; delta-type opioid receptor; DOR; D-OR-1; DOR-1; K56; mDOR; MSL-2; Nbor; Opioid receptor; opioid receptor A; opioid receptor delta 1; opioid receptor, delta 1; OPRD; Oprd1; Ror-a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100351268, 487329, 4985 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P41143 | |
OPRD1 | |
Synthetic 18 amino acid peptide from 3rd extracellular domain of human Delta Opioid Receptor. | |
50 μg | |
Primary | |
Canine, Human, Non-human Primate, Rabbit | |
Antibody | |
IgG |