missing translation for 'onlineSavingsMsg'
Learn More

Rhodopsin Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen OSR00220W

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11555103

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material.

Photoreceptor required for image-forming vision at low light intensity. Required for photoreceptor cell viability after birth. Light-induced isomerization of 11-cis to all-trans retinal triggers a conformational change leading to G-protein activation and release of all-trans retinal. Abs(max)=495 nm. Rod shaped photoreceptor cells which mediates vision in dim light. Phosphorylated on some or all of the serine and threonine residues present in the C-terminal region. Defects in RHO are the cause of retinitis pigmentosa type 4 (RP4). RP leads to degeneration of retinal photoreceptor cells. Patients typically have night vision blindness and loss of midperipheral visual field. As their condition progresses, they lose their far peripheral visual field and eventually central vision as well. RP4 inheritance is autosomal domit. Defects in RHO are also a cause of retinitis pigmentosa autosomal recessive (ARRP). Also, defects in RHO are a cause of autosomal domit congenital stationary night blindness 1 (CSNBAD1); also known as rhodopsin-related congenital stationary night blindness. Congenital stationary night blindness is a nonprogressive retinal disorder characterized by impaired night vision and ocular symptoms such as myopia, hyperopia, nystagmus and reduced visual acuity.
Spezifikation

Spezifikation

Rhodopsin
Polyclonal
RHO
Rhodopsin, opsin-2, RHO, OPN2, RP4, MGC138309, MGC138311
Rabbit
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
P08100, P15409, P51489
RHO
A synthetic peptide from c-terminal region of human Rhodopsin
RUO
212541, 24717, 6010
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote