missing translation for 'onlineSavingsMsg'
Learn More

Opsin 3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen OSR00213W

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11585093

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material.

Opsins are members of the guanine nucleotide-binding protein (G protein)-coupled receptor superfamily. In addition to the visual opsins, mammals possess several photoreceptive non-visual opsins that are expressed in extraocular tissues. This gene, opsin 3, is strongly expressed in brain and testis and weakly expressed in liver, placenta, heart, lung, skeletal muscle, kidney, and pancreas. The gene may also be expressed in the retina. The protein has the canonical features of a photoreceptive opsin protein. May play a role in encephalic photoreception. Strongly expressed in brain. Highly expressed in the preoptic area and paraventricular nucleus of the hypothalamus. Shows highly patterned expression in other regions of the brain, being enriched in selected regions of the cerebral cortex, cerebellar Purkinje cells, a subset of striatal neurons, selected thalamic nuclei, and a subset of interneurons in the ventral horn of the spinal cord.
Spezifikation

Spezifikation

Opsin 3
Polyclonal
OPN3
Opsin-3, encephalopsin, panopsin, OPN3, ECPN, opsin 3
Rabbit
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
Q9H1Y3, Q9WUK7
OPN3
Synthetic peptide from human opsin-3.
RUO
13603, 23596, 498289
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote