Learn More
Opticin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544262
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: LSDNLLDSIP GPLPLSLRSV HLQNNLIETM QRDVFCDPEE HKHTRRQLED Sequence homology: Cow: 77%; Dog: 100%; Guinea Pig: 85%; Horse: 92%; Human: 100%; Mouse: 85%; Pig: 100%; Rabbit: 92%; Rat: 100%; Zebrafish: 83%
Opticin Polyclonal antibody specifically detects Opticin in Human samples. It is validated for Western Blot.Spezifikation
Opticin | |
Polyclonal | |
Unconjugated | |
OPTC | |
dermatan sulfate proteoglycan 3; dspg3; Oculoglycan; OPT; Optc; optcl; opticin; SLRP; SLRP protein; small leucine-rich repeat protein; small leucine-rich repeat protein (SLRP); wu:fb81a05; zgc:101047 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26254 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UBM4 | |
OPTC | |
Synthetic peptide directed towards the C-terminal of human OPTC | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |