missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR10J5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA539845
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.Spezifikation
OR10J5 | |
Polyclonal | |
Unconjugated | |
OR10J5 | |
OR10J5, Olfactory receptor 10J5, OR1-28, MOR23, Olfactory receptor OR1-28 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
127385 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8NHC4 | |
OR10J5 | |
A synthetic peptide derived from the internal region of human OR10J5 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |