missing translation for 'onlineSavingsMsg'
Learn More

OR2A4 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA534049

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15372551

  • 451.00 € / 50 Mikrogramm
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Percent identity with other species by BLAST analysis: Human, Elephant, Panda, Dog (100%) Marmoset, Hamster, Bovine, Horse, Pig (92%) Mouse, Rat (85%).

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Spezifikation

Spezifikation

OR2A4
Polyclonal
Unconjugated
OR2A4
OR2A4, Olfactory receptor 2A4, Olfactory receptor 2A10, Olfactory receptor OR6-37, OR2A10
Rabbit
Antigen affinity chromatography
RUO
79541
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
O95047
OR2A4
Synthetic 13 amino acid peptide from 3rd extracellular domain of human OR2A4.
50 μg
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote