missing translation for 'onlineSavingsMsg'
Learn More

OR2D3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen OSR00277W

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11511242

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

Reconstitute with 100 μL of distilled water.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Spezifikation

Spezifikation

OR2D3
Polyclonal
OR2D3
OR2D3, OR11-89, Olfactory receptor 2D3, olfactory receptor, family 2, subfamily D, member 3
Rabbit
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
Q8NGH3
OR2D3
A synthetic peptide from human olfactory receptor
RUO
120775
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote