missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORC1 Monoclonal Antibody (TK 1/2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA190545
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-90545 detects ORC1 from Xenopus laevis samples.
ORC1 Monoclonal antibody specifically detects ORC1 in Xenopus laevis samples. It is validated for Immunoprecipitation, Western Blot.Spezifikation
ORC1 | |
Monoclonal | |
1.2 mg/mL | |
PBS with no preservative | |
0 | |
orc1.L | |
Recombinant full length protein (Xenopus laevis). | |
100 μg | |
Primary | |
X. laevis | |
Antibody | |
IgG2a |
Immunoprecipitation, Western Blot | |
TK 1/2 | |
Unconjugated | |
orc1.L | |
AA545195; HS; HSORC1; MmORC1; orc1; orc1.L; Orc1l; origin recognition complex protein 1; origin recognition complex subunit 1; origin recognition complex subunit 1 L homeolog; origin recognition complex, subunit 1; origin recognition complex, subunit 1 homolog; origin recognition complex, subunit 1-like; PARC1; Replication control protein 1; XELAEV_18023178mg; xorc1 | |
Mouse | |
Protein A | |
RUO | |
398063 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |