Learn More
ORC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA521488
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Raji. Predicted reactivity: Mouse (84%), Rat (84%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ORC2 Polyclonal antibody specifically detects ORC2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ORC2 | |
Polyclonal | |
Unconjugated | |
ORC2 | |
AU041563; Orc2; ORC2L; origin recognition complex protein 2 homolog; origin recognition complex subunit 2; origin recognition complex, subunit 2; origin recognition complex, subunit 2 homolog; origin recognition complex, subunit 2-like; zgc:153041 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4999 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.45 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q13416 | |
ORC2 | |
Recombinant protein fragment contain a sequence corresponding to a region within amino acids 208 and 479 (Q13416) of ORC2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |