missing translation for 'onlineSavingsMsg'
Learn More

ORC3 Monoclonal Antibody (1D6), Invitrogen™

Rat Monoclonal Antibody

Marke:  Invitrogen MA517024

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15328530

  • 473.00 € / 50 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

MA5-17024 reacts with ORC3 in human samples. MA5-17024 has been successfully used in immunoprecipitation and Western blot applications. The MA5-17024 immunogen is ORC3 (hsORC) produced in P3-x63Ag.8.653 cells.

ORC3 Monoclonal antibody specifically detects ORC3 in Human samples. It is validated for Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

ORC3
Monoclonal
1 mg/mL
PBS with 0.1% sodium azide
Q9UBD5
ORC3
ORC3 (hsORC) produced in P3-x63Ag.8.653 cells
50 μL
Primary
Human
Antibody
IgG2b
Immunoprecipitation, Western Blot
1D6
Unconjugated
ORC3
homolog of latheo, Drosophila; IMAGE50150; LAT; LATHEO; Orc3; ORC3L; origin of replication 3 homolog; origin recognition complex subunit 3; Origin recognition complex subunit Latheo; origin recognition complex, subunit 3; origin recognition complex, subunit 3 honolog; origin recognition complex, subunit 3-like; OTTHUMP00000040611; RP3-486L4.1
Rat
Protein G
RUO
23595
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote