Learn More
Orexin A Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Marke: Invitrogen PA19583
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-9583 detects Orexin A in human, mouse, and rat samples. PA1-9583 has been successfully used in ELISA procedures. PA1-9583 immunogen is a synthetic peptide corresponding to amino acids 1-13 (AQPLPDCCRQKTC) of human Orexin A.
Orexin A Polyclonal antibody specifically detects Orexin A in Human, Mouse, Rat samples. It is validated for ELISA.Spezifikation
Orexin A | |
Polyclonal | |
Unconjugated | |
HCRT | |
HCRT; Hcrt1; Hcrt2; Hypocretin; Hypocretin (orexin) neuropeptide; hypocretin neuropeptide precursor; hypocretin-1; Hypocretin-2; NRCLP1; orexin; orexin A; orexin B; orexin-A; Orexin-B; ORX; OX; PPORX; PPOX; prepro-orexin | |
Chicken | |
Affinity chromatography | |
RUO | |
15171, 25723, 3060 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
0.5 mg/mL | |
PBS with 0.02% sodium azide | |
O43612, O55232, O55241 | |
HCRT | |
Synthetic peptide corresponding to amino acids 1-13 of human Orexin A. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |