missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orexin Receptor 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533837
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Bovine, Rabbit, Pig, Platypus (100%).
Orexin Receptor 1 Polyclonal antibody specifically detects Orexin Receptor 1 in Bovine, Human, Rabbit, Rat samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
Orexin Receptor 1 | |
Polyclonal | |
Unconjugated | |
HCRTR1 | |
Hcrt receptor 1; Hcrtr1; Hctr1; hypocretin (orexin) receptor 1; hypocretin receptor 1; hypocretin receptor type 1; hypocretin receptor-1; Orexin a receptor; orexin receptor 1; orexin receptor type 1; orexin receptor-1; Orexin-1 receptor; OX1 receptor; OX1R; ox-1-R; ox1-R; OX-R1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100352294, 25593, 282248, 3061 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O43613, P56718, Q0GBZ5 | |
HCRTR1 | |
Synthetic 14 amino acid peptide from 2nd extracellular domain of human HCRTR1 / Orexin Receptor 1. | |
50 μg | |
Primary | |
Bovine, Human, Rabbit, Rat | |
Antibody | |
IgG |