missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORMDL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA524232
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, rat and Xenopus and zebrafish based on sequence homology.
ORMDL3 Polyclonal antibody specifically detects ORMDL3 in Human, Mouse samples. It is validated for Western Blot.Spezifikation
ORMDL3 | |
Polyclonal | |
Unconjugated | |
ORMDL3 | |
2810011N17Rik; AI315223; ORM1-like 3 (S. cerevisiae); ORM1-like protein 3; ORMDL sphingolipid biosynthesis regulator 3; Ormdl3 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
66612, 94103 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q8N138, Q9CPZ6 | |
ORMDL3 | |
KLH conjugated synthetic peptide between 53-81 amino acids from the central region of human ORMDL3 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |