Learn More
ODC1 Monoclonal Antibody (MP16-2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA513485
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-13485 targets Ornithine Decarboxylase in IHC (F) applications and shows reactivity with Human, mouse, and Rat samples. The MA5-13485 immunogen is a synthetic hexadecapeptide (KPDEKYYSSSIWGPTC) (aa345-360) from rat ODC.
ODC1 Monoclonal antibody specifically detects ODC1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen).Spezifikation
ODC1 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P00860, P09057, P11926 | |
Odc1 | |
A synthetic hexadecapeptide (KPDEKYYSSSIWGPTC) (aa345-360) from rat ODC | |
500 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Frozen) | |
MP16-2 | |
Unconjugated | |
Odc1 | |
MGC103389; Odc; ODC1; Ornithine decarboxylase; ornithine decarboxylase 1; ornithine decarboxylase, structural 1; Ornitine decarboxylase | |
Mouse | |
Protein G | |
RUO | |
18263, 24609, 4953 | |
4° C | |
Liquid |