missing translation for 'onlineSavingsMsg'
Learn More

OSTC Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA532958

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13444629

  • 473.00 € / 50 Mikrogramm
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset, Mouse, Rat, Dog, Hamster, Panda, Pig, Opossum, Chicken, Platypus, Xenopus, Pike, Catfish, Salmon, Sablefish, Pufferfish, Zebrafish, Stickleback (100%); Monkey, Bovine (93%); Elephant, Horse, Mosquito, Ant (86%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

OSTC Polyclonal antibody specifically detects OSTC in Canine, Chicken, Hamster, Human, Mouse, Porcine, Rat, Xenopus laevis samples. It is validated for Immunohistochemistry (Paraffin).
Spezifikation

Spezifikation

OSTC
Polyclonal
Unconjugated
OSTC
2310008M10Rik; 5730557H03Rik; DC2; HDCMD45P; HSPC307; Hydrophobic protein HSF-28; oligosaccharyltransferase complex non-catalytic subunit; oligosaccharyltransferase complex subunit; oligosaccharyltransferase complex subunit (non-catalytic); oligosaccharyltransferase complex subunit OSTC; Ostc; ostc {ECO:0000250; RGD1560708; UniProtKB:Q9NRP0}
Rabbit
Antigen affinity chromatography
RUO
100517665, 100753091, 362040, 422525, 477512, 58505, 66357
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
B0K025, P86218, Q5ZJR3, Q78XF5, Q9NRP0
OSTC
Synthetic 14 amino acid peptide from internal region of human OSTC.
50 μg
Primary
Canine, Chicken, Hamster, Human, Mouse, Porcine, Rat, X. laevis
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote