missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSTC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534060
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset (100%) Mouse, Rat, Hamster, Panda, Bovine, Dog, Pig, Opossum, Xenopus (94%).
OSTC Polyclonal antibody specifically detects OSTC in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
OSTC | |
Polyclonal | |
Unconjugated | |
OSTC | |
2310008M10Rik; 5730557H03Rik; DC2; HDCMD45P; HSPC307; Hydrophobic protein HSF-28; oligosaccharyltransferase complex non-catalytic subunit; oligosaccharyltransferase complex subunit; oligosaccharyltransferase complex subunit (non-catalytic); oligosaccharyltransferase complex subunit OSTC; Ostc; ostc {ECO:0000250; RGD1560708; UniProtKB:Q9NRP0} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
58505 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9NRP0 | |
OSTC | |
Synthetic 17 amino acid peptide from N-Terminus of human OSTC. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |