Learn More
OTUB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA521402
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa, HepG2, mouse brain, rat brain. Predicted reactivity: Mouse (100%), Rat (100%), Dog (99%), Pig (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
OTUB1 Polyclonal antibody specifically detects OTUB1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
OTUB1 | |
Polyclonal | |
Unconjugated | |
OTUB1 | |
AI850305; Deubiquitinating enzyme OTUB1; deubiquitinating enzyme OTUB1 {ECO:0000250; EMBL:AAI66771.1}; FLJ20113; FLJ40710; hOTU1; HSPC263; hypothetical protein LOC540984; MGC111158; MGC4584; OTB1; OTU deubiquitinase, ubiquitin aldehyde binding 1; OTU domain, ubiquitin aldehyde binding 1; OTU domain-containing ubiquitin aldehyde-binding protein 1; OTU domain-containing ubiquitin aldehyde-binding protein 1 {ECO:0000250; OTU1; OTUB1; otub1 {ECO:0000312; otubain 1; otubain-1; otubain-1 {ECO:0000250; OTU-domain Ubal-binding 1; Ubiquitin thioesterase OTUB1; ubiquitin thioesterase OTUB1 {ECO:0000250; ubiquitin-specific protease otubain 1; ubiquitin-specific-processing protease OTUB1; ubiquitin-specific-processing protease OTUB1 {ECO:0000250; UniProtKB:Q7TQI3} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
107260, 293705, 55611 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.249 mg/mL | |
PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
B2RYG6, Q7TQI3, Q96FW1 | |
OTUB1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 149 of OTUB1 (Uniprot ID#Q96FW1) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |